Please login to your online account to display your discounted pricing

IL-6 Rat anti-Human, Biotin, Clone: MQ2-39C3, eBioscience™

Rat Monoclonal Antibody

$143.55 - $361.05

Specifications

Antigen IL-6
Clone MQ2-39C3
Host Species Rat
Gene Alias Interleukin-6, IL6
Species Reactivity Human
View More Specs

 Disclaimers

For Research Use Only.

Products
Catalog Number Mfr. No. Quantity Price Quantity    

5012165

 
affymetrix
13706881
50μg Each for $143.55

5012166

 
affymetrix
13706885
500μg Each for $361.05
Description & Specifications

Specifications

Antigen IL-6
Clone MQ2-39C3
Host Species Rat
Gene Alias Interleukin-6, IL6
Species Reactivity Human
Applications ELISA Detection
Applications ELISPOT Detection Antibody
Regulatory Status RUO
Conjugate Biotin
Format Conjugated
Storage Requirements Store at 2-8°C. Do not freeze.
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Formulation aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer
Concentration 0.5mg/mL

The MQ2-39C3 antibody reacts with human interleukin-6 (IL-6), a 21-28kDa cytokine secreted by a variety of cell types.