missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

IL-6 Rat anti-Human, APC, Clone: MQ2-13A5, eBioscience™

Rat Monoclonal Antibody

$174.24 - $389.40

Specifications

Antigen IL-6
Clone MQ2-13A5
Host Species Rat
Gene Alias Interleukin-6
Species Reactivity Human
View More Specs

Disclaimers:

For Research Use Only.



Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
50245700 Life Technologies
LS17706941
25 Tests Each for $174.24
50245701 Life Technologies
LS17706942
100 Tests Each for $389.40
Specifications

Description

The MQ2-13A5 antibody reacts with human interleukin-6 (IL-6), a 21-28kDa cytokine secreted by a variety of cell types.
Description & Specifications

Specifications

IL-6
Rat
Human
RUO
Conjugated
Primary
aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer
MQ2-13A5
Interleukin-6
Flow Cytometry,Flow Cytometry (Intracellular Staining)
Allophycocyanin
Store at 2-8°C. Do not freeze. Light-sensitive material.
Monoclonal
5μL (0.06μg)/test