missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

IL-5 Rat anti-Mouse, Biotin, Clone: TRFK4, eBioscience™

Rat Monoclonal Antibody

$130.68 - $341.88

Specifications

Antigen IL-5
Clone TRFK4
Host Species Rat
Gene Alias Interleukin-5, IL5
Species Reactivity Mouse
View More Specs

Disclaimers:

For Research Use Only.



Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
5012159 Life Technologies
LS13705181
50μg Each for $130.68
5012160 Life Technologies
LS13705185
500μg Each for $341.88
Specifications

Description

The TRFK4 antibody reacts with mouse interleukin-5 (IL-5), also known as BCDF-α and eosinophil differentiation factor (EDF). This 15-20kDa cytokine is secreted by Th2 cells and exists a homodimer.
Description & Specifications

Specifications

IL-5
Rat
Mouse
RUO
Conjugated
Primary
aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer
TRFK4
Interleukin-5, IL5
ELISA Detection,ELISPOT
Biotin
Store at 2-8°C. Do not freeze.
Monoclonal
0.5mg/mL