Please login to your online account to display your discounted pricing

IL-5 Rat anti-Human, Mouse, Clone: TRFK5, Functional Grade, eBioscience™

Rat Monoclonal Antibody, Functional Grade

$129.05 - $2,463.55

Specifications

Antigen IL-5
Clone TRFK5
Host Species Rat
Gene Alias Interleukin-5, IL5
Species Reactivity Human, Mouse
View More Specs

 Disclaimers

For Research Use Only.

Products
Catalog Number Mfr. No. Quantity Price Quantity    

501122980

 
affymetrix
16705238
5mg Each for $2,463.55

5014700

 
affymetrix
16705281
50μg Each for $129.05

5014701

 
affymetrix
16705285
500μg Each for $433.55
Description & Specifications

Specifications

Antigen IL-5
Clone TRFK5
Host Species Rat
Gene Alias Interleukin-5, IL5
Species Reactivity Human, Mouse
Applications Cytokine Neutralization
Applications ELISA Capture
Applications ELISPOT
Applications Immunohistochemistry
Regulatory Status RUO
Conjugate Unlabeled
Format Purified
Storage Requirements Store at 2-8°C.
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Formulation aqueous buffer, no sodium azide
Concentration 1mg/mL

The TRFK5 antibody reacts with mouse and human interleukin-5 (IL-5). The TRFK5 antibody is a neutralizing antibody. Mouse IL-5 is a disulfide-linked homodimer, containing two 113 amino acid peptides; these resolve as 32-34kDa bands in SDS-PAGE. IL-5 is produced by T cells and has been known as eosinophil-differentiating factor (EDF), B cell growth factor II (BCGFII), and T cell-replacing factor (TRF). IL-5 induces eosinophil differentiation and promotes eosinophil survival and activation. In mice, IL-5 has been shown to stimulate B cell proliferation and antibody production.