missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

IL-5 Rat anti-Human, Mouse, Clone: TRFK5, eBioscience™

Rat Monoclonal Antibody

$117.48 - $315.48

Specifications

Antigen IL-5
Clone TRFK5
Host Species Rat
Gene Alias Interleukin-5
Species Reactivity Human, Mouse
View More Specs

Disclaimers:

For Research Use Only.



Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
5013612 Life Technologies
LS14705281
50μg Each for $117.48
5013613 Life Technologies
LS14705285
500μg Each for $315.48
Specifications

Description

The TRFK5 antibody reacts with mouse and human interleukin-5 (IL-5). The TRFK5 antibody is a neutralizing antibody. Mouse IL-5 is a disulfide-linked homodimer, containing two 113 amino acid peptides; these resolve as 32-34kDa bands in SDS-PAGE. IL-5 is produced by T cells and has been known as eosinophil-differentiating factor (EDF), B cell growth factor II (BCGFII), and T cell-replacing factor (TRF). IL-5 induces eosinophil differentiation and promotes eosinophil survival and activation. In mice, IL-5 has been shown to stimulate B cell proliferation and antibody production.
Description & Specifications

Specifications

IL-5
Rat
Human, Mouse
RUO
Purified
Primary
aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer
TRFK5
Interleukin-5
Cytokine Neutralization,ELISA Capture,ELISPOT Capture Antibody,Flow Cytometry (Intracellular Staining)
Unlabeled
Store at 2-8°C.
Monoclonal
0.5mg/mL