Please login to your online account to display your discounted pricing

IL-5 Rat anti-Human, Biotin, Clone: JES1-5A10, eBioscience™

Rat Monoclonal Antibody

$158.05 - $390.05

Specifications

Antigen IL-5
Clone JES1-5A10
Host Species Rat
Gene Alias Interleukin-5, IL5
Species Reactivity Human
View More Specs

 Disclaimers

For Research Use Only.

Products
Catalog Number Mfr. No. Quantity Price Quantity    

5012161

 
affymetrix
13705981
50μg Each for $158.05

5012162

 
affymetrix
13705985
500μg Each for $390.05
Description & Specifications

Specifications

Antigen IL-5
Clone JES1-5A10
Host Species Rat
Gene Alias Interleukin-5, IL5
Species Reactivity Human
Applications ELISA Detection
Applications ELISPOT Detection Antibody
Regulatory Status RUO
Conjugate Biotin
Format Conjugated
Storage Requirements Store at 2-8°C. Do not freeze.
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Formulation aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer
Concentration 0.5mg/mL

The JES1-5A10 antibody reacts with human interleukin-5 (IL-5), also known as BCDF-α and eosinophil differentiation factor (EDF). This 15-20kDa cytokine is secreted by T cells and exists a homodimer.