missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

IL-4 Rat anti-Mouse, PerCP-eFluor 710, Clone: 11B11, eBioscience™

Rat Monoclonal Antibody

$143.88 - $355.08

Specifications

Antigen IL-4
Clone 11B11
Host Species Rat
Gene Alias Interleukin-4, B-cell stimulatory factor 1
Species Reactivity Mouse
View More Specs

Disclaimers:

For Research Use Only.



Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
501123885 Life Technologies
LS46704180
25μg Each for $143.88
501123886 Life Technologies
LS46704182
100μg Each for $355.08
Specifications

Description

The 11B11 antibody reacts with mouse interleukin-4 (IL-4), a 14 kD cytokine. The 11B11 antibody is a neutralizing antibody. Mouse IL-4, also known as BCDF, BCGF-1 and BSF-1, is secreted by activated Th2 and natural killer T cells, and to a lesser extent by Th1 and mast cells. IL-4 is species-specific and promotes proliferation and differentiation of B cells. IL-4, in conjunction with other signals such as CD40, induces Ig isotype switching of B cells and the upregulation of surface MHC class II and CD23.
Description & Specifications

Specifications

IL-4
Rat
Mouse
RUO
Conjugated
Primary
aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer
11B11
Interleukin-4, B-cell stimulatory factor 1
Flow Cytometry (Intracellular Staining)
PerCP-eFluor 710
Store at 2-8°C. Do not freeze. Light-sensitive material.
Monoclonal
0.2mg/mL