Please login to your online account to display your discounted pricing

IL-4 Rat anti-Mouse, PE-Cy7, Clone: 11B11, eBioscience™

Rat Monoclonal Antibody

$191.40 - $487.20

Specifications

Antigen IL-4
Clone 11B11
Host Species Rat
Gene Alias Interleukin-4
Species Reactivity Mouse
View More Specs

 Disclaimers

For Research Use Only.

Products
Catalog Number Mfr. No. Quantity Price Quantity    

501123429

 
life technologies
LS25704180
25μg Each for $191.40

501123430

 
life technologies
LS25704182
100μg Each for $487.20
Description & Specifications

Specifications

Antigen IL-4
Clone 11B11
Host Species Rat
Gene Alias Interleukin-4
Species Reactivity Mouse
Applications Flow Cytometry (Intracellular Staining)
Regulatory Status RUO
Conjugate PE-Cy7
Format Conjugated
Storage Requirements Store at 2-8°C. Do not freeze. Light-sensitive material. This tandem dye is sensitive to photo-induced oxidation. Protect this vial from light during storage
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Formulation aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer
Concentration 0.2mg/mL

The 11B11 antibody reacts with mouse interleukin-4 (IL-4), a 14 kD cytokine. The 11B11 antibody is a neutralizing antibody. Mouse IL-4, also known as BCDF, BCGF-1 and BSF-1, is secreted by activated Th2 and natural killer T cells, and to a lesser extent by Th1 and mast cells. IL-4 is species-specific and promotes proliferation and differentiation of B cells. IL-4, in conjunction with other signals such as CD40, induces Ig isotype switching of B cells and the upregulation of surface MHC class II and CD23.