missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

IL-4 Rat anti-Mouse, PE, Clone: 11B11, eBioscience™

Rat Monoclonal Antibody

$130.68 - $328.68

Specifications

Antigen IL-4
Clone 11B11
Host Species Rat
Gene Alias Interleukin-4
Applications Flow Cytometry (Intracellular Staining)
View More Specs

Disclaimers:

For Research Use Only.



Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
5011128 Life Technologies
LS12704141
25 Tests Each for $130.68
501129606 Life Technologies
LS12704182
100μg Each for $328.68
501129670 Life Technologies
LS12704181
50μg Each for $200.64
Specifications

Description

The 11B11 antibody reacts with mouse interleukin-4 (IL-4), a 14 kD cytokine. The 11B11 antibody is a neutralizing antibody. Mouse IL-4, also known as BCDF, BCGF-1 and BSF-1, is secreted by activated Th2 and natural killer T cells, and to a lesser extent by Th1 and mast cells. IL-4 is species-specific and promotes proliferation and differentiation of B cells. IL-4, in conjunction with other signals such as CD40, induces Ig isotype switching of B cells and the upregulation of surface MHC class II and CD23.
Description & Specifications

Specifications

IL-4
Rat
Flow Cytometry (Intracellular Staining)
PE
Store at 2-8°C. Do not freeze. Light-sensitive material.
Monoclonal
0.2mg/mL; 5μL (0.06μg)/test
11B11
Interleukin-4
RUO
Conjugated
Primary
aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer