missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

IL-4 Rat anti-Mouse, FITC, Clone: BVD6-24G2, eBioscience™

Rat Monoclonal Antibody

$143.88 - $289.08

Specifications

Antigen IL-4
Clone BVD6-24G2
Host Species Rat
Gene Alias Interleukin-4
Species Reactivity Mouse
View More Specs

Disclaimers:

For Research Use Only.



Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
509995 Life Technologies
LS11704241
25 Tests Each for $143.88
509996 Life Technologies
LS11704281
50μg Each for $275.88
509997 Life Technologies
LS11704282
100μg Each for $289.08
Specifications

Description

The BVD6-24G2 antibody reacts with mouse interleukin-4 (IL-4), a 14kDa cytokine secreted by Th2 cells.
Description & Specifications

Specifications

IL-4
Rat
Mouse
RUO
Conjugated
Primary
aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer
BVD6-24G2
Interleukin-4
Flow Cytometry (Intracellular Staining)
FITC
Store at 2-8°C. Do not freeze. Light-sensitive material.
Monoclonal
0.5mg/mL; 5μL (0.25μg)/test