Please login to your online account to display your discounted pricing

IL-4 Rat anti-Mouse, Clone: 11B11, Functional Grade, eBioscience™

Rat Monoclonal Antibody, Functional Grade

$129.05 - $3,623.55

Specifications

Antigen IL-4
Clone 11B11
Host Species Rat
Gene Alias Interleukin-4
Species Reactivity Mouse
View More Specs

 Disclaimers

For Research Use Only.

Products
Catalog Number Mfr. No. Quantity Price Quantity    

501122976

 
life technologies
LS16704195
10mg Each for $3,623.55

5014696

 
life technologies
LS16704181
50μg Each for $129.05

5014697

 
life technologies
LS16704185
500μg Each for $433.55
Description & Specifications

Specifications

Antigen IL-4
Clone 11B11
Host Species Rat
Gene Alias Interleukin-4
Species Reactivity Mouse
Applications ELISA Capture
Applications ELISPOT
Applications Functional Assay
Regulatory Status RUO
Conjugate Unlabeled
Format Purified
Storage Requirements Store at 2-8°C.
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Formulation aqueous buffer, no sodium azide
Concentration 1mg/mL

The 11B11 antibody reacts with mouse interleukin-4 (IL-4), a 14 kD cytokine. The 11B11 antibody is a neutralizing antibody. Mouse IL-4, also known as BCDF, BCGF-1 and BSF-1, is secreted by activated Th2 and natural killer T cells, and to a lesser extent by Th1 and mast cells. IL-4 is species-specific and promotes proliferation and differentiation of B cells. IL-4, in conjunction with other signals such as CD40, induces Ig isotype switching of B cells and the upregulation of surface MHC class II and CD23.