Please login to your online account to display your discounted pricing

IL-4 Rat anti-Mouse, Biotin, Clone: BVD6-24G2, eBioscience™

Rat Monoclonal Antibody

$143.55 - $375.55

Specifications

Antigen IL-4
Clone BVD6-24G2
Host Species Rat
Gene Alias Interleukin-4
Species Reactivity Mouse
View More Specs

 Disclaimers

For Research Use Only.

Products
Catalog Number Mfr. No. Quantity Price Quantity    

5012155

 
affymetrix
13704281
50μg Each for $143.55

5012156

 
affymetrix
13704285
500μg Each for $375.55
Description & Specifications

Specifications

Antigen IL-4
Clone BVD6-24G2
Host Species Rat
Gene Alias Interleukin-4
Species Reactivity Mouse
Applications ELISA Detection
Applications ELISPOT
Regulatory Status RUO
Conjugate Biotin
Format Conjugated
Storage Requirements Store at 2-8°C. Do not freeze.
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Formulation aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer
Concentration 0.5mg/mL

The BVD6-24G2 antibody reacts with mouse interleukin-4 (IL-4), a 14kDa cytokine secreted by Th2 cells.