missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

IL-4 Rat anti-Human, Clone: MP4-25D2, Functional Grade, eBioscience™

Rat Monoclonal Antibody, Functional Grade

$130.68 - $2,374.68

Specifications

Antigen IL-4
Clone MP4-25D2
Host Species Rat
Gene Alias Interleukin-4, IL4
Species Reactivity Human
View More Specs

Disclaimers:

For Research Use Only.



Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
501122979 Life Technologies
LS16704838
5mg Each for $2,374.68
5014698 Life Technologies
LS16704881
50μg Each for $130.68
501129703 Life Technologies
LS16704885
500μg Each for $381.48
Specifications

Description

The MP4-25D2 antibody reacts with human interleukin-4 (IL-4), a 15-19kDa cytokine secreted by Th2 cells.
Description & Specifications

Specifications

IL-4
Rat
Human
RUO
Purified
Primary
aqueous buffer, no sodium azide
MP4-25D2
Interleukin-4, IL4
Cytokine Neutralization
Unlabeled
Store at 2-8°C.
Monoclonal
1mg/mL