Please login to your online account to display your discounted pricing

IL-4 Rat anti-Human, Clone: MP4-25D2, eBioscience™

Rat Monoclonal Antibody

$114.55 - $114.55

Specifications

Antigen IL-4
Clone MP4-25D2
Host Species Rat
Gene Alias Interleukin-4, IL4
Species Reactivity Human
View More Specs

 Disclaimers

For Research Use Only.

Products
Catalog Number Mfr. No. Quantity Price Quantity    

5013606

 
life technologies
LS14704881
50μg Each for $114.55
Description & Specifications

Specifications

Antigen IL-4
Clone MP4-25D2
Host Species Rat
Gene Alias Interleukin-4, IL4
Species Reactivity Human
Applications Flow Cytometry (Intracellular Staining)
Regulatory Status RUO
Conjugate Unlabeled
Format Purified
Storage Requirements Store at 2-8°C.
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Formulation aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer
Concentration 0.5mg/mL

The MP4-25D2 antibody reacts with human interleukin-4 (IL-4), a 15-19kDa cytokine secreted by Th2 cells.