Learn More
IL-4 Monoclonal Antibody (MP4-25D2), Biotin, eBioscience™, Invitrogen™
Rat Monoclonal Antibody
$166.00 - $433.00
Specifications
Antigen | IL-4 |
---|---|
Clone | MP4-25D2 |
Concentration | 0.5 mg/mL |
Applications | ELISA |
Classification | Monoclonal |
Description
Description: The MP4-25D2 antibody reacts with human interleukin-4 (IL-4), a 15-19 kDa cytokine secreted by Th2 cells. Applications Reported: The MP4-25D2 antibody has been reported for use in detection of human IL-4 by ELISA and for intracellular staining for flow cytometry. Applications Tested: The MP4-25D2 antibody has been tested as the detection antibody in a sandwich ELISA for analysis of human Interleukin-4 (IL-4) in combination with the purified 8D4-8 (14-7049) antibody for capture and recombinant human IL-4 (14-8049) as the standard. A suitable range of concentrations of this antibody for ELISA detection is 0.5-2 μg/mL. A standard curve consisting of doubling dilutions of the recombinant standard over the range of 500 pg/mL - 4 pg/mL should be included in each ELISA plate. Filtration: 0.2 μm post-manufacturing filtered.
Interleukin-4 (IL-4) is a pleiotropic, immune-modulatory cytokine produced by activated T cells, and serves as a ligand for interleukin 4 receptor. IL-4 has been found to have the broadest range of actions. The interleukin 4 receptor also binds to IL13, which may contribute to many overlapping functions of this cytokine and IL13. STAT6, a signal transducer and activator of transcription, has been shown to play an important role in mediating the immune regulatory signal of IL-4. The IL-4 gene, IL-3, IL-5, IL-13, and CSF2 form a cytokine gene cluster on chromosome 5q. In particular, IL-4 is located in close proximity to IL13 on the chromosome. Further, the IL-4 gene, IL13 and IL5 are found to be coordinated by several long-range regulatory elements over a 120 kilobase range on the chromosome. IL-4 delta 2 is a known splice variant of IL-4.Specifications
IL-4 | |
0.5 mg/mL | |
Monoclonal | |
Liquid | |
RUO | |
PBS with 0.09% sodium azide; pH 7.2 | |
B cell growth factor 1; B_cell stimulatory factor 1; B-cell growth factor 1; B-cell IgG differentiation factor; B-cell stimulatory factor 1; BCGF1; BCGF-1; binetrakin; BSF1; BSF-1; cytokine; H-IL-4; IGG1 induction factor; IL2; IL4; Il-4; Il4e12; ILN; Interleukin; interleukin 4; interleukin 4 variant 2; interleukin 4 variant IL-4delta2; interleukin 4 variant IL-4delta3; interleukin 4 variant IL-4int2A; interleukin 4 variant IL-4int3A; interleukin 4 variant IL-4int3B; Interleukin4; interleukin-4; interleukin-4 precursor; lymphocyte stimulatory factor 1; MGC79402; M-IL-4; Pitrakinra | |
Il4 | |
Primary | |
4° C, do not freeze | |
Il4 |
MP4-25D2 | |
ELISA | |
Biotin | |
Rat | |
Human | |
P05112 | |
3565 | |
IgG1 κ | |
Affinity chromatography | |
Antibody |
For Research Use Only.