missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

IL-4 Mouse anti-Rat, eFluor 660, Clone: OX81, eBioscience™

Mouse Monoclonal Antibody

$158.05 - $361.05

Specifications

Antigen IL-4
Clone OX81
Host Species Murine
Gene Alias Interleukin 4
Species Reactivity Rat
View More Specs

Disclaimers:

For Research Use Only.



Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
501124430 Life Technologies
LS50704580
25μg Each for $158.05
501124431 Life Technologies
LS50704582
100μg Each for $361.05
Specifications

Description

This OX81 monoclonal antibody reacts with rat interleukin (IL)-4. Also known as BCDF, BCGF-1 and BSF-1, IL-4 is secreted by activated Th2 and natural killer T cells. This cytokine is also produced by Th1 and mast cells albeit to a lesser extent. IL-4 is species-specific and promotes proliferation and differentiation of B cells. IL-4, in conjunction with other signals such as CD40, induces Ig isotype switching of B cells and the upregulation of MHC class II and CD23.

The OX81 antibody has been reported to exhibit neutralizing activity.
Description & Specifications

Specifications

IL-4
Murine
Rat
RUO
Conjugated
Primary
aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer
OX81
Interleukin 4
Flow Cytometry,Flow Cytometry (Intracellular Staining)
eFluor 660
Store at 2-8°C. Do not freeze. Light-sensitive material.
Monoclonal
0.2mg/mL