missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

IL-4 Mouse anti-Rat, Clone: OX81, Functional Grade, eBioscience™

Mouse Monoclonal Antibody, Functional Grade

$201.55 - $506.05

Specifications

Antigen IL-4
Clone OX81
Host Species Murine
Gene Alias Interleukin-4
Species Reactivity Rat
View More Specs

Disclaimers:

For Research Use Only.



Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
501122977 Life Technologies
LS16704582
100μg Each for $201.55
501122978 Life Technologies
LS16704585
500μg Each for $506.05
Specifications

Description

This OX81 monoclonal antibody reacts with rat interleukin (IL)-4. Also known as BCDF, BCGF-1 and BSF-1, IL-4 is secreted by activated Th2 and natural killer T cells. This cytokine is also produced by Th1 and mast cells albeit to a lesser extent. IL-4 is species-specific and promotes proliferation and differentiation of B cells. IL-4, in conjunction with other signals such as CD40, induces Ig isotype switching of B cells and the upregulation of MHC class II and CD23.

The OX81 antibody has been reported to exhibit neutralizing activity.
Description & Specifications

Specifications

IL-4
Murine
Rat
RUO
Purified
Primary
aqueous buffer, no sodium azide
OX81
Interleukin-4
Flow Cytometry (Intracellular Staining),Functional Assay,Western Blot
Unlabeled
Store at 2-8°C.
Monoclonal
1mg/mL