Please login to your online account to display your discounted pricing

IL-4 Mouse anti-Human, PE-Cy7, Clone: 8D4-8, eBioscience™

Mouse Monoclonal Antibody

$176.90 - $442.25

Specifications

Antigen IL-4
Clone 8D4-8
Host Species Mouse
Gene Alias Interleukin-4
Species Reactivity Human
View More Specs

 Disclaimers

For Research Use Only.

Products
Catalog Number Mfr. No. Quantity Price Quantity    

5015609

 
life technologies
LS25704941
25 tests Each for $176.90

5015610

 
life technologies
LS25704982
100μg Each for $442.25
Description & Specifications

Specifications

Antigen IL-4
Clone 8D4-8
Host Species Mouse
Gene Alias Interleukin-4
Species Reactivity Human
Applications Flow Cytometry (Intracellular Staining)
Regulatory Status RUO
Conjugate PE-Cy7
Format Conjugated
Storage Requirements Store at 2-8°C. Do not freeze. Light-sensitive material. This tandem dye is sensitive to photo-induced oxidation. Protect this vial from light during storage
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Formulation aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer
Concentration 0.2mg/mL; 5μL (0.125μg)/test

The 8D4-8 antibody reacts with human interleukin-4 (IL-4), a 15-19kDa cytokine secreted by Th2 cells.