Please login to your online account to display your discounted pricing

IL-4 Mouse anti-Human, Alexa Fluor 488, Clone: 8D4-8, eBioscience™

Mouse Monoclonal Antibody

$176.90 - $368.30

Specifications

Antigen IL-4
Clone 8D4-8
Host Species Murine
Gene Alias Interleukin-4
Species Reactivity Human
View More Specs

 Disclaimers

For Research Use Only.

Products
Catalog Number Mfr. No. Quantity Price Quantity    

5016792

 
life technologies
LS53704941
25 Tests Each for $176.90

5016793

 
life technologies
LS53704942
100 Tests Each for $368.30
Description & Specifications

Specifications

Antigen IL-4
Clone 8D4-8
Host Species Murine
Gene Alias Interleukin-4
Species Reactivity Human
Applications Flow Cytometry (Intracellular Staining)
Regulatory Status RUO
Conjugate Alexa Fluor 488
Format Conjugated
Storage Requirements Store at 2-8°C. Do not freeze. Light-sensitive material.
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Formulation aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer
Concentration 5μL (0.25μg)/test

The 8D4-8 antibody reacts with human interleukin-4 (IL-4), a 15-19kDa cytokine secreted by Th2 cells.