Please login to your online account to display your discounted pricing

IL-33R (ST2) Rat anti-Mouse, PerCP-eFluor 710, Clone: RMST2-33, eBioscience™

Rat Monoclonal Antibody

$187.05 - $477.05

Specifications

Antigen IL-33R (ST2)
Clone RMST2-33
Host Species Rat
Gene Alias IL-1RL1, IL-1R4, T1
Species Reactivity Mouse
View More Specs

 Disclaimers

For Research Use Only.

Products
Catalog Number Mfr. No. Quantity Price Quantity    

501123944

 
life technologies
LS46933380
25μg Each for $187.05

501123945

 
life technologies
LS46933382
100μg Each for $477.05
Description & Specifications

Specifications

Antigen IL-33R (ST2)
Clone RMST2-33
Host Species Rat
Gene Alias IL-1RL1, IL-1R4, T1
Species Reactivity Mouse
Applications Flow Cytometry
Regulatory Status RUO
Conjugate PerCP-eFluor 710
Format Conjugated
Storage Requirements Store at 2-8°C. Do not freeze. Light-sensitive material. This tandem dye is sensitive to photo-induced oxidation. Protect this vial from light during storage
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Formulation aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer
Concentration 0.2mg/mL

The RMST2-33 monoclonal antibody reacts with mouse ST2, also known as IL-33R or interleukin 1 receptor-like 1. ST2 is a member of the IL-1 receptor family. In the membrane-bound form, it consists of three extracellular immunoglobulin domains and an intracellular toll-interleukin-1 receptor domain. ST2 forms a dimer with IL-1R accessory protein in a ligand-dependent manner. IL-33/ST2 interaction induces the production of TH2 cytokines. ST2 also has a soluble isoform lacking transmembrane and intracellular toll-interleukin-1 receptor domains. It is believed that the soluble form functions as a decoy receptor that can block membrane bound IL-33/ST2 interaction. ST2 is expressed by TH2 lymphocytes, mast cells, eosinophils, basophils, innate lymphocytes, smooth muscle cells, and endothelial cells and is involved in host defense, allergy, and inflammation. IL-33/ST2 interaction has also been shown to be atheroprotective.

RMST2-33 has been reported to bind and activate ST2 in in vitro assays.