missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

IL-33R (ST2), APC, clone: RMST2-2, eBioscience™

Rat Monoclonal Antibody

Manufacturer:  Life Technologies LS17933582

 View more versions of this product

Catalog No. 501129755



Description

Description

The protein encoded by this gene is a member of the interleukin 1 receptor family. Studies of the similar gene in mouse suggested that this receptor can be induced by proinflammatory stimuli, and may be involved in the function of helper T cells. This gene, interleukin 1 receptor, type I (IL1R1), interleukin 1 receptor, type II (IL1R2) and interleukin 1 receptor-like 2 (IL1RL2) form a cytokine receptor gene cluster in a region mapped to chromosome 2q12. Alternative splicing of this gene results in multiple transcript variants.

It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest. Excitation: 633-647 nm; Emission: 660 nm; Laser: Red Laser. Filtration: 0.2 μm post-manufacturing filtered. ST2 is a member of the interleukin 1 receptor family. Studies of ST2 in mouse suggested that this receptor can be induced by proinflammatory stimuli, and may be involved in the function of helper T cells. ST2 belongs to the interleukin-1 receptor family with three Ig-like C2-type (immunoglobulin-like) domains and a TIR domain. ST2 is a receptor for interleukin-33 (IL-33) which upon stimulation recruits the signaling molecules MYD88, IRAK1, IRAK4, and TRAF6, followed by phosphorylation of MAPK3/ERK1 and/or MAPK1/ERK2, MAPK14, and MAPK8. Reports also suggest its possible role in helper T-cell function as well as in cell growth. Three isoforms of ST2 have been reported where isoform C is a membrane form and isoform B is a secreted form. ST2 may be induced by proinflammatory stimuli, and may also be found in serum of autoimmune disease. Different isoforms are restricted to different tissues while high expression of this protein is evident in kidney, lung, placenta, stomach, skeletal muscle, colon and small intestine.
Specifications

Specifications

IL-33R (ST2)
Monoclonal
0.2 mg/mL
PBS with 0.1% gelatin and 0.09% sodium azide; pH 7.2
P14719
DER4, Fit-1, Ly84, ST2L, St2, St2-rs1, T1, T1/ST2
Rat
Affinity chromatography
RUO
Antibody
Monoclonal
Mouse
Flow Cytometry
RMST2-2
APC
Il1rl1
Liquid
Il1rl1
IgG2a, kappa
100 μg
4° C, store in dark, DO NOT FREEZE!
Primary
17082
Documents

For Research Use Only.