missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

IL-28 Rat anti-Mouse, Clone: RN28, eBioscience™

Rat Monoclonal Antibody

$129.05 - $361.05

Specifications

Antigen IL-28
Clone RN28
Host Species Rat
Gene Alias Interleukin-28
Species Reactivity Mouse
View More Specs

Disclaimers:

For Research Use Only.



Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
5013673 Life Technologies
LS14728181
50μg Each for $129.05
5013674 Life Technologies
LS14728185
500μg Each for $361.05
Specifications

Description

This monoclonal RN28 antibody reacts with IL-28 (IFN lambda), a member of the IFN-lambda family which mediates the induction of anti-viral protection in a wide variety of cells.
Description & Specifications

Specifications

IL-28
Rat
Mouse
RUO
Purified
Primary
aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer
RN28
Interleukin-28
ELISA
Unlabeled
Store at 2-8°C.
Monoclonal
0.5mg/mL