missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

IL-28 Rabbit anti-Mouse, Biotin, Polyclonal, eBioscience™

Rabbit Polyclonal Antibody

$143.55 - $404.55

Specifications

Antigen IL-28
Host Species Rabbit
Gene Alias Interleukin-28
Species Reactivity Mouse
Applications ELISA
View More Specs

Disclaimers:

For Research Use Only.



Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
5012193 Life Technologies
LS13728281
50μg Each for $143.55
5012194 Life Technologies
LS13728285
500μg Each for $404.55
Specifications

Description

This polyconal Anti-Mouse IL-28 antibody reacts with mouse interleukin-28 (IL-28), a member of the IFN-lambda family which mediates the induction of anti-viral protection in a wide variety of cells.
Description & Specifications

Specifications

IL-28
Interleukin-28
ELISA
Biotin
Store at 2-8°C. Do not freeze.
Polyclonal
0.5mg/mL
Rabbit
Mouse
RUO
Conjugated
Primary
aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer