missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

IL-23 p19, Alexa Fluor 488, clone: fc23cpg, eBioscience™

Rat Monoclonal Antibody

Manufacturer:  Life Technologies LS53702382

 View more versions of this product

Catalog No. 501124611



Description

Description

The p19 shares sequence similarity with IL-6 subfamily members and is distantly related to the p35 subunit of IL-12. It shows no biological activity by itself; instead, it combines with the p40 subunit of IL-12 to form a biologically active, composite cytokine, IL-23. IL-23 shares some in vivo functions with IL-12, including the activation of the transcription factor Stat4. Similar to IL-12, human IL-23 stimulates IFN-g production and proliferation of PHA blast T cells, as well as in CD45 RO (memory) T cells. Ubiquitous transgenic expression of the IL-23 subunit p19 induces multiorgan inflammation, runting, infertility and premature death.

Applications Tested: This fc23cpg antibody has been tested by intracellular staining and flow cytometric analysis of stimulated mouse bone marrow-derived dendritic cells using the Intracellular Fixation & Permeabilization Buffer Set (cat. 88-8824) and protocol. Please refer to Best Protocols: Protocol A: Two step protocol for (cytoplasmic) intracellular proteins located under the Resources Tab online. This can be used at less than or equal to 0.125 µg per test. A test is defined as the amount (µg) of antibody that will stain a cell sample in a final volume of 100 µL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest. Excitation: 488 nm; Emission: 519 nm; Laser: Blue Laser. Filtration: 0.2 μm post-manufacturing filtered. The p19 shares sequence similarity with IL-6 subfamily members and is distantly related to the p35 subunit of IL-12. It shows no biological activity by itself; instead, it combines with the p40 subunit of IL-12 to form a biologically active, composite cytokine, IL-23. IL-23 shares some in vivo functions with IL-12, including the activation of the transcription factor Stat4. Similar to IL-12, human IL-23 stimulates IFN-g production and proliferation of PHA blast T cells, as well as in CD45 RO (memory) T cells. Ubiquitous transgenic expression of the IL-23 subunit p19 induces multiorgan inflammation, runting, infertility and premature death.
Specifications

Specifications

IL-23 p19
Monoclonal
0.5 mg/mL
PBS with 0.1% gelatin and 0.09% sodium azide; pH 7.2
Q9EQ14
IL-23, IL-23A, IL23P19, MGC79388, P19, SGRF
Rat
Affinity chromatography
RUO
Antibody
Monoclonal
Mouse
Flow Cytometry
fc23cpg
Alexa Fluor® 488
Il23a
Liquid
Il23a
IgG1, kappa
100 μg
4° C, store in dark, DO NOT FREEZE!
Primary
83430
Documents

For Research Use Only.