Please login to your online account to display your discounted pricing

IL-23 p19 Mouse anti-Human, Clone: HNU2319, Functional Grade, eBioscience™

Mouse Monoclonal Antibody, Functional Grade

$158.05 - $404.55

Specifications

Antigen IL-23 p19
Clone HNU2319
Host Species Mouse
Gene Alias Interleukin 23 p19, IL23 p19, IL23p19, IL-23p19
Species Reactivity Human
View More Specs

 Disclaimers

For Research Use Only.

Products
Catalog Number Mfr. No. Quantity Price Quantity    

5014558

 
life technologies
LS16523682
100μg Each for $158.05

5014559

 
life technologies
LS16523685
500μg Each for $404.55
Description & Specifications

Specifications

Antigen IL-23 p19
Clone HNU2319
Host Species Mouse
Gene Alias Interleukin 23 p19, IL23 p19, IL23p19, IL-23p19
Species Reactivity Human
Applications Cytokine Neutralization
Regulatory Status RUO
Conjugate Unlabeled
Format Purified
Storage Requirements Store at 2-8°C.
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Formulation aqueous buffer, no sodium azide
Concentration 1mg/mL

The HNU2319 monoclonal antibody is specific to the p19 subunit of human IL-23, a heterodimeric cytokine made up of two covalently linked subunits, p40 and p19. It is closely related to IL-12, with which it shares the p40 subunit. Dendritic cells and macrophages produce IL-23 in response to TLR2, TLR4, and TLR8 ligands, as well as the β-glucan receptor Dectin-1.

Human IL-23 induces proliferation of memory T cells and induces moderate levels of IFN-γ production by naïve and memory T cells, as compared to IL-12. In contrast, the mouse IL-23 biological activities are more distinctly different than those of mouse IL-12 suggesting the signaling or response elements (receptors) differ between species. The IL-23 receptor is also heterodimeric and shares the IL-12Rβ1 chain with IL-12, while the IL-23R chain is unique to IL-23. Signaling occurs through the Jak/STAT pathway and results in RORγt expression, which drives the differentiation of CD4+ T-cells towards the Th17 phenotype.

The monoclonal antibody HNU2319 neutralizes the bioactivity of IL-23 without affecting IL-12.