Please login to your online account to display your discounted pricing

IL-23 Mouse anti-Human, PerCP-eFluor 710, Clone: 23dcdp, eBioscience™

Mouse Monoclonal Antibody

$230.55 - $535.05

Specifications

Antigen IL-23
Clone 23dcdp
Host Species Mouse
Gene Alias Interleukin-23 p19
Species Reactivity Human
View More Specs

 Disclaimers

For Research Use Only.

Products
Catalog Number Mfr. No. Quantity Price Quantity    

501123911

 
life technologies
LS46782341
25 tests Each for $230.55

501123912

 
life technologies
LS46782342
100 tests Each for $535.05
Description & Specifications

Specifications

Antigen IL-23
Clone 23dcdp
Host Species Mouse
Gene Alias Interleukin-23 p19
Species Reactivity Human
Applications Flow Cytometry (Intracellular Staining)
Regulatory Status RUO
Conjugate PerCP-eFluor 710
Format Conjugated
Storage Requirements Store at 2-8°C. Do not freeze. Light-sensitive material. This tandem dye is sensitive to photo-induced oxidation. Protect this vial from light during storage
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Formulation aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer
Concentration 5μL (0.03μg)/test

This 23dcdp monoclonal antibody reacts with the p19 subunit of human IL-23. This heterodimeric cytokine is composed of two disulfide-linked subunits, p40 and p19. It is closely related to IL-12, with which it shares the p40 subunit. The IL-23 receptor is also heterodimeric and shares the IL-12R\beta1 chain with IL-12, while the IL-23R chain is unique to the IL-23 receptor complex. IL-23R signaling occurs through the Jak/STAT pathway and results in ROR\gammat expression, which promotes maintenance and proliferation of T helper 17 (Th17) cells. Dendritic cells and macrophages produce IL-23 in response to TLR2, TLR4, and TLR8 ligands, as well as agonists of the \beta-glucan receptor, Dectin-1. Our studies suggest donor, kinetics and type of stimulant can result in variation in IL-23 expression levels. Recent publications suggest the p19 subunit may also exist in the absence of association with p40.