Please login to your online account to display your discounted pricing

IL-23 Mouse anti-Human, eFluor 660, Clone: 23dcdp, eBioscience™

Mouse Monoclonal Antibody

$216.05 - $506.05

Specifications

Antigen IL-23
Clone 23dcdp
Host Species Murine
Gene Alias Interleukin-23 p19
Species Reactivity Human
View More Specs

 Disclaimers

For Research Use Only.

Products
Catalog Number Mfr. No. Quantity Price Quantity    

5016475

 
life technologies
LS50782341
25 tests Each for $216.05

5016476

 
life technologies
LS50782342
100 Tests Each for $506.05
Description & Specifications

Specifications

Antigen IL-23
Clone 23dcdp
Host Species Murine
Gene Alias Interleukin-23 p19
Species Reactivity Human
Applications Flow Cytometry (Intracellular Staining)
Regulatory Status RUO
Conjugate eFluor 660
Format Conjugated
Storage Requirements Store at 2-8°C. Do not freeze. Light-sensitive material.
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Formulation aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer
Concentration 5μL (0.03μg)/test

This 23dcdp monoclonal antibody reacts with the p19 subunit of human IL-23. This heterodimeric cytokine is composed of two disulfide-linked subunits, p40 and p19. It is closely related to IL-12, with which it shares the p40 subunit. The IL-23 receptor is also heterodimeric and shares the IL-12Rβ1 chain with IL-12, while the IL-23R chain is unique to the IL-23 receptor complex. IL-23R signaling occurs through the Jak/STAT pathway and results in RORγt expression, which promotes maintenance and proliferation of T helper 17 (Th17) cells.

Dendritic cells and macrophages produce high levels of p40 and low levels of IL-23 in response to TLR2, TLR4, and TLR8 ligands and agonists of the β-glucan receptor, Dectin-1. This is due to the presence of monomer and dimer forms of p40, while only a small portion associates with p19 to form IL-23. Our studies suggest donor variability, expression kinetics and type of stimulant can result in variation in IL-23 expression levels. Recent publications suggest the p19 subunit may also exist in the absence of association with p40.