missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

IL-22 Rat anti-Mouse, PerCP-eFluor 710, Clone: 1H8PWSR, eBioscience™

Rat Monoclonal Antibody

$249.48 - $487.08

Specifications

Antigen IL-22
Clone 1H8PWSR
Host Species Rat
Gene Alias Interleukin-22
Applications Flow Cytometry (Intracellular Staining)
View More Specs

Disclaimers:

For Research Use Only.



Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
501123902 Life Technologies
LS46722180
25μg Each for $249.48
5016104 Life Technologies
LS46722182
100μg Each for $487.08
Specifications

Description

The 1H8PWSR monoclonal antibody reacts with mouse interleukin(IL)-22. IL-22 is a 20kDa member of the IL-10 cytokine family that is secreted primarily by Th17, NK cells, and some other T cells. In in vitro Th17 cultures, induction of IL-22 expression is greater in response to IL-23 than IL-6 or TGF-ß, suggesting that this cytokine may be secreted by more fully differentiated Th17 cells in vivo. A heterodimer consisting of IL-10R2 and IL-22R1 serves as the receptor for IL-22. Th17-secreted IL-22 binds to the IL-22 receptor complexon target cells to induce the expression of anti-microbial peptides ß-defensin-2 and ß-defensin-3. Recently, it was demonstrated that IL-22 is able to protect hosts against bacterial infections of the lungs and gut.
Description & Specifications

Specifications

IL-22
Rat
Flow Cytometry (Intracellular Staining)
PerCP-eFluor 710
Store at 2-8°C. Do not freeze. Light-sensitive material.
Monoclonal
0.2mg/mL
1H8PWSR
Interleukin-22
RUO
Conjugated
Primary
aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer