Please login to your online account to display your discounted pricing

IL-22 Mouse anti-Human, PE, Clone: 22URTI, eBioscience™

Mouse Monoclonal Antibody

$265.35 - $442.25

Specifications

Antigen IL-22
Clone 22URTI
Host Species Mouse
Gene Alias Interleukin-22
Species Reactivity Human
View More Specs

 Disclaimers

For Research Use Only.

Products
Catalog Number Mfr. No. Quantity Price Quantity    

5011180

 
life technologies
LS12722941
25 tests Each for $265.35

501128772

 
life technologies
LS12722942
100 tests Each for $442.25
Description & Specifications

Specifications

Antigen IL-22
Clone 22URTI
Host Species Mouse
Gene Alias Interleukin-22
Species Reactivity Human
Applications Flow Cytometry (Intracellular Staining)
Regulatory Status RUO
Conjugate PE
Format Conjugated
Storage Requirements Store at 2-8°C. Do not freeze. Light-sensitive material.
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Formulation aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer
Concentration 5μL (0.06μg)/test

The 22URTI monoclonal antibody reacts with human interleukin(IL)-22. IL-22 is a 20kDa member of the IL-10 cytokine family that is secreted primarily by Th17 and NK cells. Nevertheless, other T cells have also been shown to produce IL-22. In in vitro Th17 cultures, induction of IL-22 expression is greater in response to IL-23 than IL-6 or TGF beta, suggesting that this cytokine may be secreted by more fully differentiated Th17 cells in vivo. A heterodimer consisting of IL-10R2 and IL-22R1 serves as the receptor for IL-22.