missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

IL-22 Mouse anti-Human, FITC, Clone: 22URTI, eBioscience™

Mouse Monoclonal Antibody

$265.35 - $427.75

Specifications

Antigen IL-22
Clone 22URTI
Host Species Murine
Gene Alias Interleukin-22
Species Reactivity Human
View More Specs

Disclaimers:

For Research Use Only.



Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
501122158 Life Technologies
LS11722941
25 Tests Each for $265.35
501122159 Life Technologies
LS11722942
100 Tests Each for $427.75
Specifications

Description

The 22URTI monoclonal antibody reacts with human interleukin(IL)-22. IL-22 is a 20kDa member of the IL-10 cytokine family that is secreted primarily by Th17 and NK cells. Nevertheless, other T cells have also been shown to produce IL-22. In in vitro Th17 cultures, induction of IL-22 expression is greater in response to IL-23 than IL-6 or TGF beta, suggesting that this cytokine may be secreted by more fully differentiated Th17 cells in vivo. A heterodimer consisting of IL-10R2 and IL-22R1 serves as the receptor for IL-22.
Description & Specifications

Specifications

IL-22
Murine
Human
RUO
Conjugated
Primary
aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer
22URTI
Interleukin-22
Flow Cytometry (Intracellular Staining)
FITC
Store at 2-8°C. Do not freeze. Light-sensitive material.
Monoclonal
5μL (0.125μg)/test