Please login to your online account to display your discounted pricing

IL-21 Mouse anti-Human, Clone: eBio3A3-N2 (3A3-N2), eBioscience™

Mouse Monoclonal Antibody

$114.55 - $245.05

Specifications

Antigen IL-21
Clone eBio3A3-N2 (3A3-N2)
Host Species Mouse
Gene Alias Interleukin-21, IL21
Species Reactivity Human
View More Specs

 Disclaimers

For Research Use Only.

Products
Catalog Number Mfr. No. Quantity Price Quantity    

5013662

 
life technologies
LS14721980
25μg Each for $114.55

5013663

 
life technologies
LS14721981
50μg Each for $143.55

501129102

 
life technologies
LS14721982
100μg Each for $245.05
Description & Specifications

Specifications

Antigen IL-21
Clone eBio3A3-N2 (3A3-N2)
Host Species Mouse
Gene Alias Interleukin-21, IL21
Species Reactivity Human
Applications ELISA Capture
Regulatory Status RUO
Conjugate Unlabeled
Format Purified
Storage Requirements Store at 2-8°C.
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Formulation aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer
Concentration 0.5mg/mL

The monoclonal antibody eBio3A3-N2 reacts with human IL-21, a potent stimulator of NK and cytolytic T cells. IL-21 is a 131 amino acid protein most closely related to IL-2 and IL-15. Expression is found in CD4+ cells upon activation. In addition to its effect on NK cells, IL-21 stimulates proliferation of B-cell stimulated by crosslinking of the CD40 antigen and stimulates the proliferation of bone marrow progenitor cells and the expression of the NK-cell marker CD56 in the presence of IL-15.