missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

IL-20 Receptor 2 Rat anti-Human, Mouse, Clone: 20RNTC, eBioscience™

Rat Monoclonal Antibody

$143.88 - $302.28

Specifications

Antigen IL-20 Receptor 2
Clone 20RNTC
Host Species Rat
Gene Alias Interleukin-20 Receptor
Applications Flow Cytometry,Immunocytochemistry,Western Blot
View More Specs

Disclaimers:

For Research Use Only.



Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
5012839 Life Technologies
LS14120682
100μg Each for $302.28
5012838 Life Technologies
LS14120680
25μg Each for $143.88
Specifications

Description

This 20RNTC monoclonal antibody recognizes mouse and human IL-20R2 (interleukin-20 receptor 2, IL-20Rb) in either of its two heterodimeric complexes formed by pairing with IL-20R1 or IL-22R1. IL-20R2 belongs to the class II cytokine receptor family that shares a characteristic pattern of cysteine, proline and tryptophan residues in the extracellular domain. IL-20R2 paired with IL-20R1 forms a receptor that binds IL-20, IL-19, or IL-24. IL-20R2 paired with IL22R1 forms a receptor that binds IL-20 or IL-24. The IL-20R complex signals primarily via the JAK-STAT pathway. IL-20R2 is found predominantly on cells of epithelial origin such as gut and skin and can be upregulated during inflammation. Immunofluorescent staining of A375 cells reveals a punctate staining pattern at the cell membrane, as well as diffuse, cytoplasmic staining that is likely associated with protein synthesis and transport in the endoplasmic reticulum and golgi.
Description & Specifications

Specifications

IL-20 Receptor 2
Rat
Flow Cytometry,Immunocytochemistry,Western Blot
Unlabeled
Store at 2-8°C.
Monoclonal
0.5mg/mL
20RNTC
Interleukin-20 Receptor
RUO
Purified
Primary
aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer