missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

IL-2 Rat anti-Mouse, eFluor 450, Clone: JES6-5H4, eBioscience™

Rat Monoclonal Antibody

$157.08 - $335.28

Specifications

Antigen IL-2
Clone JES6-5H4
Host Species Rat
Gene Alias Interleukin-2
Species Reactivity Mouse
View More Specs

Disclaimers:

For Research Use Only.



Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
5016393 Life Technologies
LS48702180
25μg Each for $157.08
5016394 Life Technologies
LS48702182
100μg Each for $335.28
Specifications

Description

The JES6-5H4 antibody reacts with mouse interleukin-2 (IL-2), a 17kDa T cell growth factor and a major immunoregulatory cytokine.
Description & Specifications

Specifications

IL-2
Rat
Mouse
RUO
Conjugated
Primary
aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer
JES6-5H4
Interleukin-2
Flow Cytometry (Intracellular Staining)
eFluor 450
Store at 2-8°C. Do not freeze. Light-sensitive material.
Monoclonal
0.2mg/mL