Please login to your online account to display your discounted pricing

IL-2 Rat anti-Mouse, Alexa Fluor 488, Clone: JES6-5H4, eBioscience™

Rat Monoclonal Antibody

$172.55 - $361.05

Specifications

Antigen IL-2
Clone JES6-5H4
Host Species Rat
Gene Alias Interleukin-2
Species Reactivity Mouse
View More Specs

 Disclaimers

For Research Use Only.

Products
Catalog Number Mfr. No. Quantity Price Quantity    

5016788

 
life technologies
LS53702180
25μg Each for $172.55

5016789

 
life technologies
LS53702182
100μg Each for $361.05
Description & Specifications

Specifications

Antigen IL-2
Clone JES6-5H4
Host Species Rat
Gene Alias Interleukin-2
Species Reactivity Mouse
Applications Flow Cytometry (Intracellular Staining)
Regulatory Status RUO
Conjugate Alexa Fluor 488
Format Conjugated
Storage Requirements Store at 2-8°C. Do not freeze. Light-sensitive material.
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Formulation aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer
Concentration 0.5mg/mL

The JES6-5H4 antibody reacts with mouse interleukin-2 (IL-2), a 17kDa T cell growth factor and a major immunoregulatory cytokine.