missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

IL-2 Rat anti-Human, PerCP-eFluor 710, Clone: MQ1-17H12, eBioscience™

Rat Monoclonal Antibody

$216.05 - $516.20

Specifications

Antigen IL-2
Clone MQ1-17H12
Host Species Rat
Gene Alias Interleukin-2
Species Reactivity Human
View More Specs

Disclaimers:

For Research Use Only.



Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
5016099 Life Technologies
LS46702941
25 Tests Each for $216.05
5016100 Life Technologies
LS46702942
100 Tests Each for $516.20
Specifications

Description

The MQ1-17H12 antibody reacts with human interleukin-2 (IL-2), a 17kDa T cell growth factor and a major immunoregulatory cytokine. The MQ1-17H12 antibody is a non-neutralizing antibody.
Description & Specifications

Specifications

IL-2
Rat
Human
RUO
Conjugated
Primary
aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer
MQ1-17H12
Interleukin-2
Flow Cytometry (Intracellular Staining)
PerCP-eFluor 710
Store at 2-8°C. Do not freeze. Light-sensitive material.
Monoclonal
5μL (0.06μg)/test