Please login to your online account to display your discounted pricing

IL-2 Rat anti-Human, PE, Clone: MQ1-17H12, eBioscience™

Rat Monoclonal Antibody

$129.05 - $346.55

Specifications

Antigen IL-2
Clone MQ1-17H12
Host Species Rat
Gene Alias Interleukin-2
Species Reactivity Human
View More Specs

 Disclaimers

For Research Use Only.

Products
Catalog Number Mfr. No. Quantity Price Quantity    

5011125

 
life technologies
LS12702941
25 tests Each for $129.05

5011126

 
life technologies
LS12702981
50μg Each for $201.55

5011127

 
life technologies
LS12702982
100μg Each for $346.55

501122373

 
life technologies
LS12702942
100 tests Each for $332.05
Description & Specifications

Specifications

Antigen IL-2
Clone MQ1-17H12
Host Species Rat
Gene Alias Interleukin-2
Species Reactivity Human
Applications Flow Cytometry
Applications Flow Cytometry (Intracellular Staining)
Regulatory Status RUO
Conjugate PE
Format Conjugated
Storage Requirements Store at 2-8°C. Do not freeze. Light-sensitive material.
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Formulation aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer
Concentration 0.2mg/mL; 5μL (0.125μg)/test

The MQ1-17H12 antibody reacts with human interleukin-2 (IL-2), a 17kDa T cell growth factor and a major immunoregulatory cytokine. The MQ1-17H12 antibody is a non-neutralizing antibody.