missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

IL-2 Rat anti-Human, Clone: MQ1-17H12, eBioscience™

Rat Monoclonal Antibody

$114.55 - $332.05

Specifications

Antigen IL-2
Clone MQ1-17H12
Host Species Rat
Gene Alias Interleukin-2
Species Reactivity Human
View More Specs

Disclaimers:

For Research Use Only.



Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
5013600 Life Technologies
LS14702981
50μg Each for $114.55
501129623 Life Technologies
LS14702985
500μg Each for $332.05
Specifications

Description

The MQ1-17H12 antibody reacts with human interleukin-2 (IL-2), a 17kDa T cell growth factor and a major immunoregulatory cytokine. The MQ1-17H12 antibody is a non-neutralizing antibody.
Description & Specifications

Specifications

IL-2
Rat
Human
RUO
Purified
Primary
aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer
MQ1-17H12
Interleukin-2
ELISA Capture,Flow Cytometry (Intracellular Staining)
Unlabeled
Store at 2-8°C.
Monoclonal
0.5mg/mL