missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

IL-2 Rat anti-Human, APC, Clone: MQ1-17H12, eBioscience™

Rat Monoclonal Antibody

$159.50 - $390.05

Specifications

Antigen IL-2
Clone MQ1-17H12
Host Species Rat
Gene Alias Interleukin-2
Species Reactivity Human
View More Specs

Disclaimers:

For Research Use Only.



Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
5015299 Life Technologies
LS17702982
100μg Each for $390.05
5015298 Life Technologies
LS17702941
25 Tests Each for $159.50
Specifications

Description

The MQ1-17H12 antibody reacts with human interleukin-2 (IL-2), a 17kDa T cell growth factor and a major immunoregulatory cytokine. The MQ1-17H12 antibody is a non-neutralizing antibody.
Description & Specifications

Specifications

IL-2
Rat
Human
RUO
Conjugated
Primary
aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer
MQ1-17H12
Interleukin-2
Flow Cytometry (Intracellular Staining)
Allophycocyanin
Store at 2-8°C. Do not freeze. Light-sensitive material.
Monoclonal
0.2mg/mL; 5μL (0.125μg)/test