Please login to your online account to display your discounted pricing

IL-17FF (homodimer) Mouse anti-Human, eFluor 660, Clone: WU24A4P, eBioscience™

Mouse Monoclonal Antibody

$433.55 - $433.55

Specifications

Antigen IL-17FF (homodimer)
Clone WU24A4P
Host Species Mouse
Gene Alias Interleukin-17F
Species Reactivity Human
View More Specs

 Disclaimers

For Research Use Only.

Products
Catalog Number Mfr. No. Quantity Price Quantity    

501124507

 
life technologies
LS50917842
100 tests Each for $433.55
Description & Specifications

Specifications

Antigen IL-17FF (homodimer)
Clone WU24A4P
Host Species Mouse
Gene Alias Interleukin-17F
Species Reactivity Human
Applications Flow Cytometry (Intracellular Staining)
Regulatory Status RUO
Conjugate eFluor 660
Format Conjugated
Storage Requirements Store at 2-8°C. Do not freeze. Light-sensitive material.
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Formulation aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer
Concentration 5μL (0.25μg)/test

The WU24A4P monoclonal antibody reacts with human interleukin (IL)-17F homodimer (IL-17FF). This antibody specifically reacts with this homodimer and has no reactivity for the IL-17AF heterodimer. IL-17FF is a 34kDa member of the IL-17 family, which includes IL-17A, B, C, D, and E (IL-25). Within this family, IL-17A and IL-17F are produced preferentially by T helper 17 (Th17) cells, with other members produced more widely. IL-17F and IL-17A have many overlapping functions, but knockout studies have shown each to have independent functions as well. In vitro cultured Th17 cells have been shown to express IL-17FF and IL-17AA homodimers and IL-17AF heterodimers depending on the culture conditions and their differentiation state. Together, these three dimers signal through the IL-17RA/IL-17RC receptor complex to mediate immune responses at mucosal interfaces and are found at lesion sites in inflammatory bowel disease, asthma, atopic dermatitis and rheumatoid arthritis.

While other clones with specificity for IL-17F, such as SHLR17, react with the IL-17F monomer in the context both IL-17FF and IL-17AF dimers, clone WU24A4P reacts specifically with IL-17FF homodimer and does not detect IL-17AF heterodimer.