missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

IL-17F Rat anti-Human, PE-Cy7, Clone: SHLR17, eBioscience™

Rat Monoclonal Antibody

$170.28 - $434.28

Specifications

Antigen IL-17F
Clone SHLR17
Host Species Rat
Gene Alias Interleukin-17F
Applications Flow Cytometry (Intracellular Staining)
View More Specs

Disclaimers:

For Research Use Only.



Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
50245753 Life Technologies
LS25716941
25 Tests Each for $170.28
50245754 Life Technologies
LS25716942
100 Tests Each for $434.28
Specifications

Description

The SHLR17 monoclonal antibody reacts with human interleukin (IL)-17F. IL-17F is a homodimeric, 34kDa member of the IL-17 family, which includes IL-17(A), B, C, D, E (IL-25). Within this family, IL-17A and IL-17F are produced preferentially by T helper 17 (Th17) cells, with other members produced more widely. IL-17F and IL-17A have many overlapping functions, but knockout studies have shown each to have independent functions as well. \i{In vitro\i} cultured Th17 cells have been shown to express IL-17F and IL-17A homodimers and IL-17A/F heterodimers depending on the culture conditions and their differentiation state. A heterodimer of IL-17RA and IL-17RC has been shown to bind IL-17F, resulting in target cell-secretion of pro-inflammatory cytokines and chemokines, as well as neutrophil recruitment.
Description & Specifications

Specifications

IL-17F
Rat
Flow Cytometry (Intracellular Staining)
PE-Cy7
Store at 2-8°C. Do not freeze. Light-sensitive material. This tandem dye is sensitive to photo-induced oxidation. Protect this vial from light during storage
Monoclonal
5μL (0.25μg)/test
SHLR17
Interleukin-17F
RUO
Conjugated
Primary
aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer