missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

IL-17F Mouse anti-Human, Clone: H17F10A7, eBioscience™

Mouse Monoclonal Antibody

$170.28 - $368.28

Specifications

Antigen IL-17F
Clone H17F10A7
Host Species Murine
Gene Alias Interleukin-17F, IL17F
Species Reactivity Human
View More Specs

Disclaimers:

For Research Use Only.



Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
5013703 Life Technologies
LS14747980
25μg Each for $170.28
5013704 Life Technologies
LS14747982
100μg Each for $368.28
Specifications

Description

The monoclonal antibody H17F10A7 reacts with human IL-17F, a member of the IL-17 family of pro-inflammatory cytokines. Like IL-17A, IL-17F is a disulfide-linked, homodimeric glycoprotein. The IL-17F homodimer includes a classical cysteine knot motif, which is found also in the TGF-ß, BMP, and NGF superfamilies. The presence of the cysteine knot motif suggested the possibility of a heterodimeric structure, as was reported for TGF-ß and inhibin/activin. Recent reports confirm that co-expression of IL-17F and IL-17A in HEK293 cells results in the formation of biologically active IL-17F/IL-17A heterodimers, in addition to the IL-17F homodimers and IL-17A homodimers. Moreover, activated human CD4+ T cells were found to produce the IL-17A/F heterodimer, along with the corresponding homodimers. In comparing the relative potency of IL-17A, IL-17F, and IL-17A/F, all three were found to induce GRO-a secretion; IL-17A was most potent, followed by IL-17A/F heterodimer, then IL-17F (100fold lower than IL-17A). These heterodimers can be detected by immunoprecipitation with eBio64CAP17 anti-IL-17A monoclonal antibody followed by immunoblot with H17F10A7.
Description & Specifications

Specifications

IL-17F
Murine
Human
RUO
Purified
Primary
aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer
H17F10A7
Interleukin-17F, IL17F
Western Blot
Unlabeled
Store at 2-8°C.
Monoclonal
0.5mg/mL