missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

IL-17A Rat anti-Mouse, Rat, FITC, Clone: eBio17B7, eBioscience™

Rat Monoclonal Antibody

$132.00 - $174.24

Specifications

Antigen IL-17A
Clone eBio17B7
Host Species Rat
Gene Alias Interleukin-17A, Cytotoxic T-lymphocyte-associated antigen 8
Species Reactivity Mouse, Rat
View More Specs

Disclaimers:

For Research Use Only.



Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
5010017 Life Technologies
LS11717780
25μg Each for $132.00
501129414 Life Technologies
LS11717781
50μg Each for $174.24
Specifications

Description

The eBio17B7 antibody reacts with mouse and rat IL-17A with no recognition of IL-17F. Interleukin-17A (IL-17A) is a CD4+ T cell-derived cytokine that promotes inflammatory responses in cell lines and is elevated in rheumatoid arthritis, asthma, multiple sclerosis, psoriasis, and transplant rejection. The cDNA encoding human IL-17A was isolated from a library of CD4+ T cells; the encoded protein exhibits 72 percent amino acid identity with HVS13, an open reading frame from a T lymphotropic Herpesvirus saimiri, and 63 percent with mouse CTLA-8 (cytotoxic T-lymphocyte associated antigen-8). Human IL-17A exists as glycosylated 20-30 kD homodimers. High levels of IL-17A homodimer are produced by activated peripheral blood CD4+ T-cells. IL-17A enhances expression of the intracellular adhesion molecule-1 (ICAM-1) in human fibroblasts. Human IL-17A also stimulates epithelial, endothelial, or fibroblastic cells to secrete IL-6, IL-8, G-CSF, and PGE2. In the presence of human IL-17A, fibroblasts can sustain the proliferation of CD34+ hematopoietic progenitors and induce maturation into neutrophils. Mouse, rat, and human IL-17A can induce IL-6 secretion in mouse stromal cells, indicating that all homologs can recognize the mouse IL-17A receptor.

IL-23-dependent, IL-17A-producing CD4+ T cells (Th-17 cells) have been identified as a unique subset of Th cells that develops along a pathway that is distinct from the Th1- and Th2- cell differentiation pathways. The hallmark effector molecules of Th1 and Th2 cells, e.g., IFN gamma and IL-4, have each been found to negatively regulate the generation of these Th-17 cells.
Description & Specifications

Specifications

IL-17A
Rat
Mouse, Rat
RUO
Conjugated
Primary
aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer
eBio17B7
Interleukin-17A, Cytotoxic T-lymphocyte-associated antigen 8
Flow Cytometry (Intracellular Staining)
FITC
Store at 2-8°C. Do not freeze. Light-sensitive material.
Monoclonal
0.5mg/mL