missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

IL-13 Rat anti-Mouse, PerCP-eFluor 710, Clone: eBio13A, eBioscience™

Rat Monoclonal Antibody

$147.84 - $376.20

Specifications

Antigen IL-13
Clone eBio13A
Host Species Rat
Gene Alias Interleukin-13
Species Reactivity Mouse
View More Specs

Disclaimers:

For Research Use Only.



Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
501123896 Life Technologies
LS46713380
25μg Each for $147.84
501123897 Life Technologies
LS46713382
100μg Each for $376.20
Specifications

Description

The eBio13A antibody reacts with mouse IL-13. IL-13 is a cytokine produced mainly by Th2 cells, but also by antigen-primed CD8 T cells. IL-13 has a strong involvement in allergic inflammation and parasitic clearing and in cancer models has been shown to have either inhibitory or stimulatory activity depending on the tumor. In humans, IL-13 is found to play a role in isotype switching in B cells. IL-13 is implicating in down modulating macrophage activity, through the reduction of pro-inflammatory cytokines (IL-1, IL-6, IL-8, IL-10, IL-12)
Description & Specifications

Specifications

IL-13
Rat
Mouse
RUO
Conjugated
Primary
aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer
eBio13A
Interleukin-13
Flow Cytometry (Intracellular Staining)
PerCP-eFluor 710
Store at 2-8°C. Do not freeze. Light-sensitive material.
Monoclonal
0.2mg/mL