Learn More
IL-13 Monoclonal Antibody (eBio13A), PE, eBioscience™, Invitrogen™
Rat Monoclonal Antibody
$164.00 - $442.00
Specifications
Antigen | IL-13 |
---|---|
Clone | eBio13A |
Applications | Flow Cytometry |
Classification | Monoclonal |
Conjugate | PE |
Description
Description: The eBio13A antibody reacts with mouse IL-13. IL-13 is a cytokine produced mainly by Th2 cells, but also by antigen-primed CD8 T cells. IL-13 has a strong involvement in allergic inflammation and parasitic clearing and in cancer models has been shown to have either inhibitory or stimulatory activity depending on the tumor. In humans, IL-13 is found to play a role in isotype switching in B cells. IL-13 is implicating in down modulating macrophage activity, through the reduction of pro-inflammatory cytokines (IL-1, IL-6, IL-8, IL-10, IL-12) Applications Reported: The eBio13A antibody has been reported useful for ELISA and ELISPOT capture, as well as intracellular staining for flow cytometric analysis. Applications Tested: This eBio13A antibody is tested intracellular staining of cultured mouse splenocytes. It is offered in 2 formats: - μg size: has been tested intracellular staining of cultured mouse splenocytes. This can be used at less than or equal to 0.25 μg per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest. - test size: has been pre-titrated and tested intracellular staining of cultured mouse splenocytes. This can be used at 5 μL (0.125 μg) per test.
IL-13 is an immunoregulatory cytokine produced primarily by activated Th2 cells, and is involved in regulating inflammatory and immune responses. IL-13 down-regulates macrophage activity, thereby inhibiting the production of pro-inflammatory cytokines and chemokines, and is involved in allergen-induced asthma through mechanisms independent of IgE and eosinophils. IL-13 also regulates several stages of B cell maturation and differentiation, up-regulates CD23 and MHC class II expression, and promotes IgE isotype switching of B cells. Structurally, IL-13 closely resembles IL-3, IL-4, IL-5 and GM-CSF. The gene encoding IL-13, IL3, IL5, IL4, and CSF2 form a cytokine gene cluster on chromosome 5q, with the IL-13 gene particularly close to IL4.Specifications
IL-13 | |
Flow Cytometry | |
PE | |
Rat | |
Mouse | |
ALRH; ALRH antibody; BHR1; il 13; Il13; Il-13; IL-13; cytokine; ILN; Interleukin; interleukin 13; Interleukin13; interleukin-13; interleukin-13 precursor; L13; MGC116786; MGC116788; MGC116789; NC30; NC30 antibody; P600; P600 antibody; T-cell activation protein P600 | |
Il13 | |
Primary | |
4° C, store in dark, DO NOT FREEZE! | |
Il13 |
eBio13A | |
Monoclonal | |
Liquid | |
RUO | |
P20109 | |
16163 | |
IgG1 κ | |
Affinity chromatography | |
Antibody |
For Research Use Only.