Please login to your online account to display your discounted pricing

IL-13 Mouse, PE, Clone: 85BRD, eBioscience™

Mouse Monoclonal Antibody

$187.05 - $419.05

Specifications

Antigen IL-13
Clone 85BRD
Host Species Murine
Gene Alias Interleukin-13, NC30
Species Reactivity Human
View More Specs

 Disclaimers

For Research Use Only.

Products
Catalog Number Mfr. No. Quantity Price Quantity    

501122379

 
life technologies
LS12713641
25 Tests Each for $187.05

501122380

 
life technologies
LS12713642
100 Tests Each for $419.05
Description & Specifications

Specifications

Antigen IL-13
Clone 85BRD
Host Species Murine
Gene Alias Interleukin-13, NC30
Species Reactivity Human
Applications Flow Cytometry (Intracellular Staining)
Regulatory Status RUO
Conjugate PE
Format Conjugated
Storage Requirements Store at 2-8°C. Do not freeze. Light-sensitive material.
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Formulation aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer
Concentration 5μL (0.125μg)/test

The 85BRD monoclonal antibody reacts with human interleukin-13 (IL-13). IL-13, also known as NC30, is a 12.5kDa protein secreted by activated T helper cells, CD8+ T cells, and NK cells. The biological activities of IL-13 include suppression of macrophage cytotoxic activity, upregulation of IL-1RA expression and suppression of inflammatory cytokine expression.