Please login to your online account to display your discounted pricing

IL-12/IL-23 Rat anti-Mouse, eFluor 660, Clone: C17.8, eBioscience™

Rat Monoclonal Antibody

$201.55 - $506.05

Specifications

Antigen IL-12/IL-23
Clone C17.8
Host Species Rat
Gene Alias Interleukin-12, IL12 / Interleukin-23, IL23 p40
Species Reactivity Mouse
View More Specs

 Disclaimers

For Research Use Only.

Products
Catalog Number Mfr. No. Quantity Price Quantity    

501124436

 
life technologies
LS50712380
25μg Each for $201.55

501124437

 
life technologies
LS50712382
100μg Each for $506.05
Description & Specifications

Specifications

Antigen IL-12/IL-23
Clone C17.8
Host Species Rat
Gene Alias Interleukin-12, IL12 / Interleukin-23, IL23 p40
Species Reactivity Mouse
Applications Flow Cytometry (Intracellular Staining)
Regulatory Status RUO
Conjugate eFluor 660
Format Conjugated
Storage Requirements Store at 2-8°C. Do not freeze. Light-sensitive material.
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Formulation aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer
Concentration 0.2mg/mL

The C17.8 antibody reacts with the p40 subunit of IL-12 and IL-23. The C17.8 antibody has been found to neutralize IL-12 and IL-23 bioactivity. For specific neutralization of IL-23 only, please see clone G23-8 (anti-mouse IL-23 p19). IL-12 is a 70kDa heterodimeric cytokine composed of two covalently linked chains, p40 and p35, and is secreted by activated monocytes, macrophages, and dendritic cells. IL-23 is a heterodimer composed of the disulfide-linked p19 protein and p40 protein, expressed by activated dendritic cells.