Please login to your online account to display your discounted pricing

IL-12/IL-23 Rat anti-Mouse, Clone: C15.6, eBioscience™

Rat Monoclonal Antibody

$129.05 - $346.55

Specifications

Antigen IL-12/IL-23
Clone C15.6
Host Species Rat
Gene Alias Interleukin-12, IL12) / IL-23 (Interleukin-23, IL23) p40
Species Reactivity Mouse
View More Specs

 Disclaimers

For Research Use Only.

Products
Catalog Number Mfr. No. Quantity Price Quantity    

5013636

 
life technologies
LS14712581
50μg Each for $129.05

5013637

 
life technologies
LS14712585
500μg Each for $346.55
Description & Specifications

Specifications

Antigen IL-12/IL-23
Clone C15.6
Host Species Rat
Gene Alias Interleukin-12, IL12) / IL-23 (Interleukin-23, IL23) p40
Species Reactivity Mouse
Applications ELISA Capture
Regulatory Status RUO
Conjugate Unlabeled
Format Purified
Storage Requirements Store at 2-8°C.
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Formulation aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer
Concentration 0.5mg/mL

The C15.6 antibody reacts with the p40 subunit of mouse interleukin-12 and IL-23. IL-12 is a 70kDa heterodimeric cytokine composed of two covalently linked chains, p40 and p35, and is secreted by activated B cells and macrophages. IL-23 is a structurally and biologically-related cytokine comprised of the same p40 subunit and a p19 subunit.

For optimal detection of total IL-12 p40 (monomer, heterodimer, homodimer) by ELISA, a combination of purified C15.6 antibody for capture and biotinylated C17.8 antibody for detection is recommended. Fluorochrome conjugated C15.6 antibody is effective for staining intracellular IL-12.