missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

IL-12/IL-23 Rat anti-Mouse, Biotin, Clone: C17.8, eBioscience™

Rat Monoclonal Antibody

$130.68 - $368.28

Specifications

Antigen IL-12/IL-23
Clone C17.8
Host Species Rat
Gene Alias Interleukin-12, IL12 / Interleukin-23, IL23 p40
Applications ELISA Detection
View More Specs

Disclaimers:

For Research Use Only.



Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
5012177 Life Technologies
LS13712381
50μg Each for $130.68
5012178 Life Technologies
LS13712385
500μg Each for $368.28
Specifications

Description

The C17.8 antibody reacts with the p40 subunit of IL-12 and IL-23. The C17.8 antibody has been found to neutralize IL-12 and IL-23 bioactivity. For specific neutralization of IL-23 only, please see clone G23-8 (anti-mouse IL-23 p19). IL-12 is a 70kDa heterodimeric cytokine composed of two covalently linked chains, p40 and p35, and is secreted by activated monocytes, macrophages, and dendritic cells. IL-23 is a heterodimer composed of the disulfide-linked p19 protein and p40 protein, expressed by activated dendritic cells.
Description & Specifications

Specifications

IL-12/IL-23
Rat
ELISA Detection
Biotin
Store at 2-8°C. Do not freeze.
Monoclonal
0.5mg/mL
C17.8
Interleukin-12, IL12 / Interleukin-23, IL23 p40
RUO
Conjugated
Primary
aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer