missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

IL-12/IL-23 Mouse anti-Human, PE, Clone: C8.6, eBioscience™

Mouse Monoclonal Antibody

$196.68 - $328.68

Specifications

Antigen IL-12/IL-23
Clone C8.6
Host Species Murine
Gene Alias Interleukin-12/Interleukin-23 p40/70
Species Reactivity Human
View More Specs

Disclaimers:

For Research Use Only.



Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
5011156 Life Technologies
LS12712981
50μg Each for $196.68
5011157 Life Technologies
LS12712982
100μg Each for $328.68
Specifications

Description

The C8.6 antibody reacts with the p40 subunit of human IL-12 p70 and human IL-23. IL-12 is a 70kDa heterodimeric cytokine composed of two covalently linked chains, p40 and p35, and secreted primarily by activated monocytes, macrophages, and dendritic cells. IL-23 is a biologically-related, heterodimeric cytokine composed of disulfide-linked p40 and p19 subunits.
Description & Specifications

Specifications

IL-12/IL-23
Murine
Human
RUO
Conjugated
Primary
aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer
C8.6
Interleukin-12/Interleukin-23 p40/70
Flow Cytometry (Intracellular Staining)
PE
Store at 2-8°C. Do not freeze. Light-sensitive material.
Monoclonal
0.2mg/mL; 5μL (0.06μg)/test