Please login to your online account to display your discounted pricing

IL-12/IL-23 Mouse anti-Human, eFluor 660, Clone: C8.6, eBioscience™

Mouse Monoclonal Antibody

$172.55 - $433.55

Specifications

Antigen IL-12/IL-23
Clone C8.6
Host Species Mouse
Gene Alias Interleukin-12/Interleukin-23, p40/70
Species Reactivity Human
View More Specs

 Disclaimers

For Research Use Only.

Products
Catalog Number Mfr. No. Quantity Price Quantity    

501124438

 
life technologies
LS50712941
25 tests Each for $172.55

501124439

 
life technologies
LS50712942
100 tests Each for $433.55
Description & Specifications

Specifications

Antigen IL-12/IL-23
Clone C8.6
Host Species Mouse
Gene Alias Interleukin-12/Interleukin-23, p40/70
Species Reactivity Human
Applications Flow Cytometry (Intracellular Staining)
Regulatory Status RUO
Conjugate eFluor 660
Format Conjugated
Storage Requirements Store at 2-8°C. Do not freeze. Light-sensitive material.
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Formulation aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer
Concentration 5μL (0.03μg)/test

The C8.6 antibody reacts with the p40 subunit of human IL-12 p70 and human IL-23. IL-12 is a 70kDa heterodimeric cytokine composed of two covalently linked chains, p40 and p35, and secreted primarily by activated monocytes, macrophages, and dendritic cells. IL-23 is a biologically-related, heterodimeric cytokine composed of disulfide-linked p40 and p19 subunits.